Foam and TPE gaskets
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SF500-FOAM S2000-FOAM TCCSFI
MMMASF500-FOAMEA 5790 DKK
MMMASF500-FOAM MMMAS2000-FOAM MMMATCCSFI
Foam and TPE gaskets
Briller
Foam and TPE gaskets are a range of protective inserts designed for 3M™ safety glasses.
  • Insert fills the gap between the eyewear and the wearers face to increase protection levels
  • Foam gasket provides improved mechanical impact absorption
  • Foam gasket also offers extra comfort and protection from sweat
  • TPE gasket (with or without rx-prescription insert) available to protect against liquids and large dust particles
  • Designed to fit specific ranges of 3M™ safety glasses
Reduce the risk of injury from projectiles and other debris with 3M™ foam and TPEMore Product Information