Scotch-Brite™ 2010 High Performance Microfibre Wipes
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2010HLWS 2010HLGN2 SB2010YE SB2010HPW
MMMA2010HLWSEA 0 DKK
MMMA2010HLWS MMMA2010HLGN2 MMMASB2010YE MMMASB2010HPW
Scotch-Brite™ 2010 High Performance Microfibre Wipes
Papirservietter Genanvendelige rengøringsklude Klude til følsomme opgaver
The 2010 series Scotch-Brite™ high performance microfibre wipes offer highly efficient, lint-free cleaning on virtually any surface, even delicate ones.
  • Highly efficient, lint-free cleaning on virtually any surface
  • Can be used with or without water, and with or without detergent
  • Extremely durable, wipes can resist up to 500 wash cycles
  • Machine washable up to 95 °C
Easily remove dust, dirt, oil, and grease with Scotch-Brite™ High Performance Microfibre Wipes 2010 Series. These high performance microfibre wipes offer highly efficient lint-free cleaning on virtually any surface, even delicate ones. Wipes are streak-free, and can be used with or without water, and with or without detergent. They’re also extremely durable, and can resist up to 500 washes at 95 °C.