Powered air respirator radiant heat covers, Versaflo™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
M-973 M-975
MMMAM-973EA 6030 DKK
MMMAM-973 MMMAM-975
Powered air respirator radiant heat covers, Versaflo™
Åndedrætsværn Åndedrætsbeskyttelsessystemer
These Versaflo™ Radiant Heat Covers are made from lightweight, aluminised aramid fabric and provide improved head, neck, and shoulder protection and comfort from radiant heat. The covers are designed for use with Versaflo™ helmets M-307 and M-407.
  • Extend M-Series use in high temperature environments
  • Lightweight aluminised aramid fabric
  • CE Approved
Extend the use of M-307 and M-407 helmets in high temperature environments with Versaflo™ Radiant Heat Covers.