Essential microfibre wipes, 2012 series, Scotch-Brite™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2012GN 2012BU 2012GB 2012RT
MMMA2012GNEA 700 DKK
MMMA2012GN MMMA2012BU MMMA2012GB MMMA2012RT
Essential microfibre wipes, 2012 series, Scotch-Brite™
Papirservietter Genanvendelige rengøringsklude Klude til alment brug
The 2012 series Scotch-Brite™ essential microfibre wipes 2012 are cleaning wipes that efficiently remove oil, grease, dirt, water, and dust from all common cleaning surfaces, even delicate ones such as mirror and marble.
  • Microfibre wipe for everyday use
  • Economical cleaning wipe with a soft touch on hands
  • Eliminates the need for cleaning agents in many cases
  • Machine washable up to 200 times at 95 °C
For everyday cleaning of common surfaces, use Scotch-Brite™ Essential Microfibre Wipes, 2012 series. These microfibre cleaning wipes can be used on allMore Product Information