Slip-resistant coarse tape, 700 series, Safety-Walk™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
AR4SZ25 AR4SZ50 AR4SZ100 SW4SC122 SW4SC914N SW4SC25 SW4SC336 SW4SC50 SW4SC150 SW4SC610 SW4SC100 SW4SC305
MMMAAR4SZ25EA 5490 DKK
MMMAAR4SZ25 MMMAAR4SZ50 MMMAAR4SZ100 MMMASW4SC122 MMMASW4SC914N MMMASW4SC25 MMMASW4SC336 MMMASW4SC50 MMMASW4SC150 MMMASW4SC610 MMMASW4SC100 MMMASW4SC305
Slip-resistant coarse tape, 700 series, Safety-Walk™
Tape
3M™ Safety-Walk™ Slip Resistant Coarse 700 Series tape provides additional slip resistance in extreme, heavy traffic areas. It contains extra large mineral particles that help provide a stable slip-resistant surface even when oil and mud are present.
  • Anti-slip tape for extreme and heavy traffic areas
  • Extra-large abrasive particles resist clogging in muddy or oily conditions
  • Can be painted without losing friction properties
  • High performance adhesive
The open abrasive pattern resists clogging and reduces clean-up time. Suitable for use on agricultureMore Product Information