Slip-resistant conformable tape, 500 series, Safety-Walk™
Leverandør: 3M
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
SW2SC50 SW2SC150 SW2SC305 SW2SC100 SW2GB50
MMMASW2SC50EA 2690 DKK
MMMASW2SC50 MMMASW2SC150 MMMASW2SC305 MMMASW2SC100 MMMASW2GB50
Slip-resistant conformable tape, 500 series, Safety-Walk™
Tape
3M™ Safety-Walk™ Slip-Resistant Conformable Tape 500 Series is a high friction, slip-resistant material that has a special aluminium backing to conform around edges and adhere to irregular surfaces.
  • Anti-slip tape for irregular or contoured surfaces in light to heavy shoe-traffic areas
  • Suitable for application in dry, wet, or oily environments
  • Aggressive, high performance adhesive forms an excellent bond for a secure hold
  • Tapes and treads resist wrinkling, curling, tearing, shrinking, and lifting
Primarily for use on dimpled or patterned surfaces inMore Product Information