Accessories for refrigerated and heating circulators, MAGIO™ MS Series
Leverandør: JULABO GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
9900100 9970245 9810007 8901120
JULA9900100EA 3690 DKK
JULA9900100 JULA9970245 JULA9810007 JULA8901120
Accessories for refrigerated and heating circulators, MAGIO™ MS Series
Cirkulatorsystemer Åbne cirkulatorsystemer
The Booster Heater offers the option of increasing the heating capacity of some MAGIO laboratory circulators by 6 kW, thereby extending their operative range. The Booster Heater can be used in a temperature range from -50 to 300 °C. Intelligent control in combination with MAGIO ensures that it can be integrated into fully automated systems.
  • The Booster Heater consists of two components:
  • Control unit
  • Heater unit with 2×3 kW
  • The heater unit is inserted into the bath opening instead of the bath lid. The control unit can be placed on or next to the circulator