ProBox storage boxes
Leverandør: MULTIROIR
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
2773 2776 2777 2784 2778 2779 2780
216-2367EA 432 DKK
216-2367 216-2368 216-2369 216-2373 216-2370 216-2371 216-2372
ProBox storage boxes
Bokse Opbevaringsbokse
These storages boxes are a premium organising system developed with a vision of functionality, high quality, and a clear Scandinavian design.
  • Made of translucent material PP (polypropylene)
  • Stackable
  • Guaranteed food-safe
  • Hygienically flawless and easy to clean
  • Ergonomic handles
  • Suitable for –40 to 120 °C storage
  • 100% recyclable
The volume of the boxes ranges between 3 and 53 L, which makes the range suitable for a wide range of storage needs. All boxes are modular and stackable for optimal space-saving storage. The recess and furrows in the lidMore Product Information