PFA Adapter Fittings, Compression to Male Threaded, Straight
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31321-43 31321-45 31321-62 31321-63
MFLX31321-43EA 963 DKK
MFLX31321-43 MFLX31321-45 MFLX31321-62 MFLX31321-63
PFA Adapter Fittings, Compression to Male Threaded, Straight
Fittings og stik
Superior chemical resistance with durable PFA.
  • Excellent choice to connect rigid tubing systems
  • Ideal for general-purpose and single-use applications
These PFA compression to male thread adapter fittings are ideal for use in corrosive environments and chemical applications. Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.