Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
68561-42 68561-43 68561-47 68561-48 68561-49 68561-51 68561-53 68561-55 68561-57 68561-59 68561-60 68561-66 68561-68 68561-69
MFLX68561-44EA 515 DKK
MFLX68561-44 MFLX68561-50 MFLX68561-52 MFLX68561-56 MFLX68561-58 MFLX68561-61 MFLX68561-62 MFLX68561-63 MFLX68561-64 MFLX68561-65 MFLX68561-67 MFLX68561-42 MFLX68561-43 MFLX68561-47 MFLX68561-48 MFLX68561-49 MFLX68561-51 MFLX68561-53 MFLX68561-55 MFLX68561-57 MFLX68561-59 MFLX68561-60 MFLX68561-66 MFLX68561-68 MFLX68561-69
Masterflex® Direct-Reading Variable-Area Flowmeters for Helium, Acrylic, Avantor®
Flow-metre
Designed for labs and plant processes where multiple gases are used.
  • Easy to read English or metric scales
  • Interchangeable scales for routine gases
  • Customer convertible panel to partial or full in-line mounted configurations
  • Panel mounting by means of hex nuts
  • Easy disassembly and assembly for cleaning
Versatile acrylic flowmeters are designed specifically for helium. Flowmeters are offered with or without built-in low hysteresis needle valves. Panel mount flowmeters without valves or use in partial or full in-line mounted configurations. FlowmetersMore Product Information