Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PFA, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06383-91 06383-92 06383-93
MFLX06383-91EA 366 DKK
MFLX06383-91 MFLX06383-92 MFLX06383-93
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, PFA, Avantor®
Fittings og stik
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.