Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
31701-82 31702-04 31702-08
MFLX30401-72EA 180 DKK
MFLX30401-72 MFLX30615-05 MFLX30615-09 MFLX30621-12 MFLX30621-14 MFLX30621-16 MFLX30621-18 MFLX30621-20 MFLX30621-26 MFLX30621-31 MFLX30621-35 MFLX30621-42 MFLX30621-46 MFLX30621-48 MFLX30621-52 MFLX30621-54 MFLX30621-56 MFLX30621-58 MFLX30621-62 MFLX30621-64 MFLX30621-66 MFLX30621-70 MFLX30621-73 MFLX30621-75 MFLX30621-77 MFLX30625-31 MFLX30625-35 MFLX30625-42 MFLX30625-46 MFLX30625-52 MFLX30625-54 MFLX30625-56 MFLX30625-64 MFLX30625-73 MFLX30625-75 MFLX30625-77 MFLX31701-82 MFLX31702-04 MFLX31702-08 MFLX30401-73 MFLX30621-08 MFLX30621-38 MFLX30625-26 MFLX30625-58 MFLX30625-62 MFLX30625-70 MFLX31517-18 MFLX31517-21
Masterflex® Adapter Fittings, Hose Barb to Male NPT Threaded, Straight, HDPE, Avantor®
Fittings og stik
Quick and easy assembly.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb