Masterflex® Union Fittings, Hose Barb, Straight, Nylon, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX30621-82EA 238 DKK
MFLX30621-82 MFLX30621-86 MFLX30621-90 MFLX30621-93 MFLX30622-56 MFLX30625-86 MFLX30625-88 MFLX30625-90 MFLX31518-06 MFLX40612-06 MFLX40622-45 MFLX40622-48 MFLX40622-54 MFLX40626-45 MFLX41517-94 MFLX30621-80 MFLX30621-84 MFLX30621-88 MFLX30622-51 MFLX30626-51 MFLX30626-56 MFLX31518-08 MFLX31518-11 MFLX40626-48 MFLX41518-03
Masterflex® Union Fittings, Hose Barb, Straight, Nylon, Avantor®
Fittings og stik
Quick and easy assembly without tools.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Chemical resistant
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.