Bürkle Cylindrical Barbed Tee Connectors, PVDF
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06044-08 06044-09 06044-11 06044-12
MFLX06044-08EA 110 DKK
MFLX06044-08 MFLX06044-09 MFLX06044-11 MFLX06044-12
Bürkle Cylindrical Barbed Tee Connectors, PVDF
Fittings og stik
Safe and flexible connection for your most demanding applications.
  • Withstands temperatures up to 320 °F (160 °C)
  • Highly resistant to chemicals and UV
  • Autoclavable
The cylindrical barbed tee connectors are available in a variety of sizes to extend, connect, branch, join, and repair most plastic tubings. The polyvinylidene fluoride (PVDF) connectors are designed for quick and easy assembly without any tools.