Masterflex® Concentric Reducer Fittings, Sanitary Tri-Clamp, Straight, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
40618-15 40618-16
MFLX31200-31EA 791 DKK
MFLX31200-31 MFLX31200-32 MFLX40618-17 MFLX40618-15 MFLX40618-16
Masterflex® Concentric Reducer Fittings, Sanitary Tri-Clamp, Straight, Avantor®
Fittings og stik
Maintain sanitary conditions.
  • Ideal for general-purpose and single-use applications, to connect tubing with rest of system
  • Determine which connector type is needed for your application by contacting free technical application assistance*
  • Global market leader in range of materials for most extensive chemical compatibility
  • Single-use custom configuration solutions to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
  • Meet