Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX96102-17EA 1680 DKK
MFLX96102-17 MFLX96102-18 MFLX96102-24 MFLX96102-35 MFLX96102-36 MFLX96103-14 MFLX96103-15 MFLX96103-16 MFLX96103-17 MFLX96103-18 MFLX96103-24 MFLX96103-25 MFLX96103-36 MFLX96102-13 MFLX96102-14 MFLX96102-15 MFLX96102-16 MFLX96102-25 MFLX96103-13
Masterflex® L/S® Sanitary Fitting Pump Tubing Assembly, PharMed® BPT, Avantor®
Slanger Peristaltisk pumpeslange
1/2" mini-clamp sanitary fittings are molded onto each end of the assembly for easy implementation into your high-purity system.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Cell and tissue culture work, pharmaceutical, light-sensitive samples
Characteristics: Nontoxic, nonhemolytic, extremely low permeability, resists acids, bases, and oxidizing agents; heat-sealable, bondable, formable, opaque beige
Requires sanitary gaskets for both connections to ensure quality seal
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.