Masterflex® Extension Cables for Modular Drives, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07557-51EA 4260 DKK
MFLX07557-51 MFLX77301-82
Masterflex® Extension Cables for Modular Drives, Avantor®
Kabler
Increase modular drive flexibility with extended cable lengths.
  • Easily connect to your Masterflex® pump with connectors on both ends
Masterflex extension cables can be used with L/S and I/P modular drives to extend the distance between drive motor and controller by 9 ft (2,7 m) to increase installation flexibility. Do not extend more than 25 ft.