Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
45520-01 45520-02 45520-03 45520-05 45520-06 45520-11 45520-12 45520-13 45520-15 45520-16 45520-21 45520-22 45520-23 45520-25 45520-26 45520-31 45520-32 45520-33 45520-35 45520-36 45520-41 45520-42 45520-43 45520-45 45520-46 45520-53 45520-55 45520-56
MFLX45520-01EA 209 DKK
MFLX45520-01 MFLX45520-02 MFLX45520-03 MFLX45520-05 MFLX45520-06 MFLX45520-11 MFLX45520-12 MFLX45520-13 MFLX45520-15 MFLX45520-16 MFLX45520-21 MFLX45520-22 MFLX45520-23 MFLX45520-25 MFLX45520-26 MFLX45520-31 MFLX45520-32 MFLX45520-33 MFLX45520-35 MFLX45520-36 MFLX45520-41 MFLX45520-42 MFLX45520-43 MFLX45520-45 MFLX45520-46 MFLX45520-53 MFLX45520-55 MFLX45520-56
Masterflex® Color-Coded Luer Fittings, Polyolefin, Avantor®
Fittings og stik
Quick and easy assembly without tools.
  • Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.