Masterflex® L/S® Precision Pump Tubing, Tygon® E-Food, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX06418-13EA 626 DKK
MFLX06418-13 MFLX06418-14 MFLX06418-16 MFLX06418-17 MFLX06418-18 MFLX06418-25
Masterflex® L/S® Precision Pump Tubing, Tygon® E-Food, Avantor®
Slanger Peristaltisk pumpeslange
Designed specifically for handling food products.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Food and beverage processing or dispensing
Characteristics: Nonwetting, odorless, tasteless, nontoxic, transparent
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.