Masterflex® Transfer Tubing, Tygon® 2375, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX95630-10EA 1020 DKK
MFLX95630-10 MFLX95630-11 MFLX95630-12 MFLX95630-13 MFLX95630-14 MFLX95630-15 MFLX95630-16 MFLX95630-17 MFLX95630-20 MFLX95630-21 MFLX95630-22 MFLX95630-23 MFLX95630-18 MFLX95630-19
Masterflex® Transfer Tubing, Tygon® 2375, Avantor®
Slanger Overførselsslange
Won't allow particulate build-up.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
These transparent tubes are ideal for use with highly aggressive chemicals – acids, bases, ketones, salts, and alcohols. Characteristics include plasticizer free, smoother inner surface, hydrophobic, minimal outgassing, safeMore Product Information