CPC® Plastic Quick-Disconnect Fittings, Sealing Plugs
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX31311-68EA 166 DKK
MFLX31311-68 MFLX31311-78 MFLX31311-69 MFLX31311-68BLK MFLX31311-78BLK MFLX31311-69BLK
CPC® Plastic Quick-Disconnect Fittings, Sealing Plugs
Fittings og stik
Fittings connect in a snap.
  • Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
  • Polycarbonate and polysulfone materials meet USP Class VI requirements
These CPC® plastic quick-disconnect fittings feature an ergonomic design and thumb latch for an easy grip and simple operation. Simply plug the coupling insert into the coupling body to connect fittings. Fittings will lock into place immediately. To disconnect fitting halves, simply press the button on the coupling body.
A complete coupler requires a fitting body and an insert (order separately).