Masterflex® Transfer Tubing, High-Purity PFA-450, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
06368-57 06368-60
MFLX06368-50EA 866 DKK
MFLX06368-50 MFLX06368-51 MFLX06368-52 MFLX06368-53 MFLX06368-54 MFLX06368-55 MFLX06368-56 MFLX06368-58 MFLX06368-59 MFLX06368-57 MFLX06368-60
Masterflex® Transfer Tubing, High-Purity PFA-450, Avantor®
Slanger Overførselsslange
Won't crack under stress.
  • Ideal for general-purpose, single-use, and extended fluid path applications
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
Applications: Pharmaceutical, biotechnology, environmental, semiconductor processing, food and beverage, autoanalyzer, and nuclear applications.
Characteristics: Superior surface smoothness and clarity, low extractables with deionized water and acids, high resistance to stress and cracking. Translucent.
Resin certification: Resin is FDA compliant.
Tubing is manufactured according to GMP.
Temperature range: -320 to 500 °F (-195 to 260 °C)
Sterilize: With autoclave, ethylene oxide, gamma irradiation, and using dry heat.