Hedland Flow Meters for Air and Other Compressed Gases
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32510-57 32510-58 32510-78 32510-80 32510-59
MFLX32510-57EA 3770 DKK
MFLX32510-57 MFLX32510-58 MFLX32510-78 MFLX32510-80 MFLX32510-59
Hedland Flow Meters for Air and Other Compressed Gases
Flow-metre
Direct read 360° rotatable scale.
  • Easy-to-read linear scale
FEATURES
Direct readings
Install in any position
360° Rotatable Guard/Scale
No Flow Straighteners or Special Piping Required
Insensitive to Shock and Vibration
Good Viscosity Stability
Temperature up to 240 °F
Accuracy +/-2% Full Scale
Repeatability +/-1%
Special Scales Available