Masterflex® L/S® Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX06442-13EA 2000 DKK
MFLX06442-13 MFLX06442-14 MFLX06442-16 MFLX06442-17 MFLX06442-18 MFLX06442-25
Masterflex® L/S® Precision Pump Tubing, Chem-Durance® Bio, Avantor®
Slanger Peristaltisk pumpeslange
Excellent chemical resistance and long pumping life.
  • Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Unique packaging options include packs, cases, and spools for customer-friendly ordering and delivery
Applications: Laboratory, pharmaceutical processing, and biotech
Characteristics: Excellent chemical resistance, excellent life and durability under pressure, low spallation, plasticizer-free inner liner with elastomer jacket, opaque
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.