Masterflex® Ismatec® Microbore Pump Tubing, PharMed® BPT, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX95809-10EA 2000 DKK
MFLX95809-10 MFLX95809-12 MFLX95809-14 MFLX95809-18 MFLX95809-22 MFLX95809-24 MFLX95809-26 MFLX95809-28 MFLX95809-30 MFLX95809-32 MFLX95809-34 MFLX95809-36 MFLX95809-38 MFLX95809-40 MFLX95809-42 MFLX95809-44 MFLX95809-46 MFLX95809-48
Masterflex® Ismatec® Microbore Pump Tubing, PharMed® BPT, Avantor®
Slanger Peristaltisk pumpeslange
Protect your light-sensitive samples.
  • Tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Regulatory approvals to match your unique application
The overall length of PharMed® BPT 95809-series extension tubing is 100 feet (30 m). The legacy Ismatec extension tubing was 3 m long.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.