Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
3401 3402 3412 3413 3414 3415 3420 3421 3422 3428 7910
734-1563EA 3580 DKK
734-1563 734-1564 734-1567 734-1568 734-1569 734-1570 734-1572 734-1573 734-1574 734-1576 CORN7910
Corning® Permeable Supports, Polycarbonate (PC) Membrane, Corning
Mikroplader Indsatsplader
75 mm Transwell® with 0,4 µm pore polycarbonate membrane insert, sterile.
  • 10 µm thick translucent polycarbonate membrane
  • Treated for optimal cell attachment
  • Each insert is packaged in a 100 mm dish, 12 inserts per case
  • Membrane must be stained for cell visibility
Produces a cell culture environment that closely resembles an in vivo state. An essential tool for the study of both anchorage-dependent and independent cell lines.