RPMI 1640, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
50-020-PC 50-020-PB 90-022-PB
50-020-PCEA 578 DKK
50-020-PC 50-020-PB 90-022-PB
RPMI 1640, Corning®
Celledyrkningsmedier
Named for the site at which it was developed, the Roswell Park Memorial Institute, RPMI 1640 is a modification of McCoy's 5A containing an increased level of inositol.
  • Low endotoxin - < 0,25 EU/ml
  • Mycoplasma tested
  • Liquid media products are formulated with cell culture grade WFI water
  • Manufactured in a cGMP environment
RPMI 1640 is also used for suspension cultures or monolayer human leukemic cells. Examples of cell types for which this medium is appropriate include mammalian myeloma and hybridoma cells such as mouse hybridomas, human leukocytes, and B and T lymphocytes.