Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
32447-00 32447-02 32447-10 32447-30 32447-34 32447-40 32447-42 32447-50 32447-54 32447-60 32447-62
MFLX32447-20EA 4640 DKK
MFLX32447-20 MFLX32447-32 MFLX32447-52 MFLX32447-00 MFLX32447-02 MFLX32447-10 MFLX32447-30 MFLX32447-34 MFLX32447-40 MFLX32447-42 MFLX32447-50 MFLX32447-54 MFLX32447-60 MFLX32447-62
Masterflex® Direct-Reading, Variable-Area Flowmeters, Stainless Steel Housing, Avantor®
Flow-metre
Ideal for the most aggressive process applications.
  • Thick polycarbonate front shield provides protection
  • Unique design facilitates ease of maintenance
  • Easy-to-read direct-reading scale offers both metric and English measurements
  • Available in fluted or plain tapered tubes
Detachable, clear 3/16"-thick polycarbonate front shield provides protection at maximum rated temperature and pressure. Easily read the graduated direct-reading scale. The interchangeable transparent scale plates mount directly on the front of the flowtube. Unique float stops allowMore Product Information