Manual Tubing Pinch Valves, Delrin®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
98002-00 98002-10 98002-20
MFLX98002-00EA 441 DKK
MFLX98002-00 MFLX98002-10 MFLX98002-20
Manual Tubing Pinch Valves, Delrin®
Ventiler Magnetventiler
An ideal option to secure tubing.
  • Constructed of durable Delrin®
Fluid contacts only the tubing—ideal for use in pharmaceutical and food industries where eliminating contamination is critical. Obtain precise and repeatable flow rates using easy-to-read metered scale on valve body. Valves are made of Delrin®. Tubing is not included—order flexible tubing with durometer (Shore A) of 60 or less from our "Tubing" section.