Hose Clamps, Adjustable, Stainless Steel
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX06403-11EA 179 DKK
MFLX06403-11 MFLX06403-12 MFLX06403-13 MFLX06403-14 MFLX06403-15 MFLX06403-16 MFLX06403-21 MFLX06403-22 MFLX06403-23 MFLX06403-24 MFLX06403-25
Hose Clamps, Adjustable, Stainless Steel
Klemmer Rørklemmer
Ideal for securely clamping tubes in place.
  • Resists corrosion
The band-type worm drive clamp features stainless steel strap and screw housing with a stainless steel screw.