Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX95714-10EA 1050 DKK
MFLX95714-10 MFLX95714-12 MFLX95714-14 MFLX95714-18 MFLX95714-22 MFLX95714-24 MFLX95714-26 MFLX95714-28 MFLX95714-30 MFLX95714-32 MFLX95714-34 MFLX95714-36 MFLX95714-38 MFLX95714-40 MFLX95714-42 MFLX95714-44 MFLX95714-46 MFLX95714-48
Masterflex® Ismatec® Pump Tubing, 3-Stop Microbore, PharMed® BPT, Avantor®
Slanger Peristaltisk pumpeslange
Designed for optimal operation in your Ismatec pumps.
  • Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
  • Global market leader in range of formulations for most extensive chemical compatibility
  • Single-use custom lengths and configurations available to minimise downtime
  • Regulatory approvals to match your unique application
  • Recommended for cell cultures and tissues
  • Impermeable to normal light and UV-radiation
To complete your Ismatec® pumping system, order three-stop color-coded tubing and extension tubing. Tubing comes in various formulations for almost any pumping application.
Three pre-attached tubing stops give you two sections—when one section of tubing fatigues, a fresh section is ready to use. The stops on the three-stop tubing are color-coded according to tubing ID. Use three-stop tubing with any of the Ismatec® minicartridge pumps and pump heads. Minicartridges withstand pressures up to 22 psi.
The overall length of PharMed BPT 95714-series stop tubing is 16" (406 mm). The legacy Ismatec® stop tubing was 400 mm long.
Order extension tubing under "Accessories". For technical assistance selecting tubing formulations and sizes, contact our Application Specialists.