Masterflex® Ismatec® Peristaltic Pump Footswitches, Avantor®
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MFLX07610-25EA 9630 DKK
MFLX07610-25 MFLX78003-07 MFLX78016-93 MFLX78016-91 MFLX78016-95
Masterflex® Ismatec® Peristaltic Pump Footswitches, Avantor®
Pumper Slangepumper
Simplify your dispensing.
  • Select the footswitch with connectio suitable for your Ismatec® pump
Footswitches provide convenient, hands-free operation of your pump drives from up to six feet away. Simply press once to start pump, press again to stop.