GF Signet Metal Flowmeter Installation Tee Fittings
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05615-26 05615-27 05620-41 05620-43 05620-45 05620-46 05620-56
MFLX05615-26EA 3310 DKK
MFLX05615-26 MFLX05615-27 MFLX05620-41 MFLX05620-43 MFLX05620-45 MFLX05620-46 MFLX05620-56
GF Signet Metal Flowmeter Installation Tee Fittings
Flow-metre
Ensure proper insertion depth and measurement.
  • Choose from 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper tee fittings
These metal installation tee fittings are available in 316 Stainless Steel (SS), carbon steel (CS), galvanized iron (schedule 40), and copper materials in a wide range of sizes from 3/4” to 8” pipe inner diameters.
Standard metal tees constructed of SS, CS, and galvanized iron fittings include PVDF insert for all sizes and NPT(F) threads at each end. Copper tee fittings include PVDF insert for pipe ID over 1" and feature sweat-on ends.
Weldolet CS fittings weld directly onto hole cut in pipe.