GF Signet Installation Tee Fittings, CPVC/PVC 80
Leverandør: Avantor Fluid Handling
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
05620-21 05620-23 05620-25 05620-26 05620-29 05620-33
MFLX05620-21EA 1690 DKK
MFLX05620-21 MFLX05620-23 MFLX05620-25 MFLX05620-26 MFLX05620-29 MFLX05620-33
GF Signet Installation Tee Fittings, CPVC/PVC 80
Flow-metre
Easily install your flowmeters.
  • Constructed of durable CPVC/PVC 80
PVC Tee Fittings for PVC 80 pipes up to 4" in diameter and CPVC 80 pipes up to 1 1/2" in diameter. Slip on ends (no threads) sensors.