PCR tube strips and dome cap strips, Axygen®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
PCR-0208-C-100 PCR-02CP-C-100
AXYGPCR-0208-C-100EA 1010 DKK
AXYGPCR-0208-C-100 AXYGPCR-02CP-C-100
PCR tube strips and dome cap strips, Axygen®
Rør PCR rør
These polypropylene tube strips and cap strips are specifically designed for PCR use and are ideal for thermal transfer applications.
  • Made from highly polished molds
  • Free of lubricants, dyes, heavy metals, and fillers
The ultra-thin and consistent tube walls ensure that thermal transfer is precisely even along the surface of the tube. The unique cap design guarantees a perfect fit to prevent sample evaporation during thermal cycling. Tubes offer a maximum RCF of 4000 g. Tubes and caps are certified by lot number to be RNase-, DNase-, and pyrogen-free.