Single-use bags for HYPERStack® vessels
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
91-200-75 91-200-76 91-200-77
MDTC91-200-75EA 1540 DKK
MDTC91-200-75 MDTC91-200-76 MDTC91-200-77
Single-use bags for HYPERStack® vessels
Poser Medieposer
These single-use bags can be connected to tubing via tube welding or by using the pre-assembled multipurpose connectors.
They can be used to add media, trypsin, or quenching substrates to complete cell culture in an entirely closed solution.