1536-Well cyclic olefin copolymer microplates, Corning®
Leverandør: Corning
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4371 4372 4370 4373 4675 4676 4677 4705
CORN4371EA 9590 DKK
CORN4371 CORN4372 CORN4370 CORN4373 CORN4675 CORN4676 CORN4677 CORN4705
1536-Well cyclic olefin copolymer microplates, Corning®
Mikroplader HTS mikroplader
1536-Well Cyclic Olefin Copolymer microplates are perfectly suited for high-throughput screening (HTS).
  • Bar coded on the short side of the microplate
  • Cyclic Olefin Copolymer (COC) material
  • Flat top for improved sealing
  • Ultra clear 127 µm film thickness
  • Variety of treatment options
  • Kalypsys and GNF system compatible
The ultraclear, 127 μm film bottom thickness is ideal for cell-based assays that require advanced optical analysis. The COC microplates are mechanically stable and inherently resistant to a broad range of solvents including DMSO and alcohols, allowing for compound storage as well.
A variety of surface treatments enable both cell-based and biochemical assays.