VWR®, Septa ND13
Leverandør: VWR Collection
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
548-0483AEA 3450 DKK
548-0483A 548-0793A 548-0487A 548-2415A 548-0482A 548-0486A 548-0484A
VWR®, Septa ND13
Septa
Almost all septa have one laminated side with PTFE (high chemical resistance) and form an inert barrier between carrier material and sample.
  • Fits for a broad range of caps
  • Easy exchange
  • Different hardness and septa thickness available
Septa choice depends on the application.