ESD safety shoes, OWEN, low, S2
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
729470/40 729470/41 729470/42 729470/43 729470/44 729470/45 729470/46 729470/47 729470/48
ELTN729470/40EA 738 DKK
ELTN729470/40 ELTN729470/41 ELTN729470/42 ELTN729470/43 ELTN729470/44 ELTN729470/45 ELTN729470/46 ELTN729470/47 ELTN729470/48
ESD safety shoes, OWEN, low, S2
Sko og støvler
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
  • Hydrophobised microfibre/mesh material
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD PRO black
  • ESD soft-fleece insole with integrated omissions
  • TPU/PU sole WELLMAXX
  • Composite toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF. Leather-free.