ESD safety shoes, LEONARDO XXSG, low, S3
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
728711/36 728711/37 728711/38 728711/39 728711/40 728711/41 728711/42 728711/43 728711/44 728711/45 728711/46 728711/47 728711/48
ELTN728711/36EA 567 DKK
ELTN728711/36 ELTN728711/37 ELTN728711/38 ELTN728711/39 ELTN728711/40 ELTN728711/41 ELTN728711/42 ELTN728711/43 ELTN728711/44 ELTN728711/45 ELTN728711/46 ELTN728711/47 ELTN728711/48
ESD safety shoes, LEONARDO XXSG, low, S3
Sko og støvler
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
  • Cowhide leather
  • Fabric lining for natural ventilation
  • Full, padded bellows tongue
  • Full-length inlay sole ARTISAN ESD
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole WELLMAXX Safety Grip
  • PU toe protection
  • Steel toe cap
Excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.