safety shoes, RENEGADE Work GTX, low, S3 CI
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5612/39 5612/40 5612/41 5612/42 5612/43 5612/44 5612/45 5612/46 5612/47
114-9050EA 1140 DKK
114-9050 114-9051 114-9052 114-9053 114-9054 114-9055 114-9056 114-9057 114-9058
safety shoes, RENEGADE Work GTX, low, S3 CI
Sko og støvler
Excellent wearing comfort, with a padded upper edge to protect the Achilles tendon.
  • Hydrophobized nubuck leather/Hydrophobized textile material
  • GORE-TEX Performance Comfort Footwear
  • Full, padded bellows tongue
  • Full-length inlay sole LOWA
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole VIBRAM® with MONOWRAP®-technology
  • Steel toe cap
The tongue prevents pressure marks and dirt from entering the shoe. The heel loop makes it easier to get inside the shoe.