ESD safety ankle boots, TERENCE XXG PRO GTX, Mid, S3 HI CI
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
768651/39 768651/40 768651/41 768651/42 768651/43 768651/44 768651/45 768651/46 768651/47 768651/48
ELTN768651/39EA 1080 DKK
ELTN768651/39 ELTN768651/40 ELTN768651/41 ELTN768651/42 ELTN768651/43 ELTN768651/44 ELTN768651/45 ELTN768651/46 ELTN768651/47 ELTN768651/48
ESD safety ankle boots, TERENCE XXG PRO GTX, Mid, S3 HI CI
Sko og støvler
Excellent wearing comfort, with an ankle-wrapping, softly padded upper edge that provides stability and grip.
  • Cowhide leather
  • GORE-TEX Performance Comfort Footwear
  • Full, padded bellows tongue
  • Full-length inlay sole ESD PRO black
  • Non-metallic, penetration-resistant midsole
  • Rubber/PU sole WELLMAXX Grip
  • TPU scuff cap and composite toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Sole core made of Infinergy® by BASF.
Certificeringer: EN ISO 20345 S3 HI, CI, HRO, SRC