ESD safety shoes, lace-up, jo_FUN Low, Jori
Leverandør: ELTEN
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
12711/40 12711/41 12711/42 12711/43 12711/44 12711/45 12711/46 12711/47 12711/48 12701/40 12701/41 12701/42 12701/43 12701/44 12701/45 12701/46 12701/47 12701/48
ELTN12711/40EA 440 DKK
ELTN12711/40 ELTN12711/41 ELTN12711/42 ELTN12711/43 ELTN12711/44 ELTN12711/45 ELTN12711/46 ELTN12711/47 ELTN12711/48 ELTN12701/40 ELTN12701/41 ELTN12701/42 ELTN12701/43 ELTN12701/44 ELTN12701/45 ELTN12701/46 ELTN12701/47 ELTN12701/48
ESD safety shoes, lace-up, jo_FUN Low, Jori
Sko og støvler
Excellent wearing comfort, with a padded upper edge that protects the Achilles tendon.
  • Hydrophobised polyester CORDURA®
  • Fabric lining for natural ventilation
  • Padded tongue
  • Full-length inlay sole ESD
  • Non-metallic, penetration-resistant midsole
  • PU/PU sole jo_TIGHT
  • PU toe protection and steel toe cap
Thanks to its excellent discharge capability, the shoe is suitable for work in ESD sensitive or electrostatically protected areas (EPA). Leather-free.
Certificeringer: EN ISO 20345 S3 SRC, form A