Cleanroom tables with perforated top for optimum airflow
Leverandør: TEKNOMEK
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
MTSSPU1200600S TSSP1200X600 TSSP1800X900/4
TEKMMTSSPU1200600SEA 0 DKK
TEKMMTSSPU1200600S TEKMTSSP1200X600 TEKMTSSP1800X900/4
Cleanroom tables with perforated top for optimum airflow
Møbler Borde
This perforated workbench is perfect for any environment where airflow is critical, the perforations allow air to flow through the hygienic stainless steel top and optional shelf.
  • 304-grade stainless steel
  • Supplied fully welded, no construction or assembly
  • Brushed dull polish finish
  • Perforated top, 6 mm diameter holes
  • Optional perforated shelf
  • Static or mobile - BZP or stainless steel castors
304 grade stainless steel, 6 mm diameter perforations, 40 mm box section frame.
Load capacity: 225 kg (for models with castors) and 250 kg (for models with feet).