Nylon cloth labels for M210, M210-LAB, and M211 label printers
Leverandør: Brady
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
152090 152091 152094 152092 152093
BRDY152090EA 240 DKK
BRDY152090 BRDY152091 BRDY152094 BRDY152092 BRDY152093
Nylon cloth labels for M210, M210-LAB, and M211 label printers
Etiketter
Nylon cloth with permanent adhesive allows labelling in environments with heat, cold, oil, and dirt. Ideal for wire and cable marking, electronic component marking, laboratory vial identification, and general labelling.
  • Permanent acrylic adhesive
  • Rounded corners
  • Suitable for freezer, liquid nitrogen, and autoclave
  • Service temperature: −40 to 90 °C
  • For use with M210 and M211 portable label printers
Aggressive permanent adhesive is suitable for low temperature use. Nylon cloth labels offer abrasion resistance, fuel/oil resistance, and high adhesion.More Product Information