Bowl, stainless steel, deep form
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
4208-0040 4208-0041 4208-0042
BURK4208-0040EA 950 DKK
BURK4208-0040 BURK4208-0041 BURK4208-0042
Bowl, stainless steel, deep form
Bakker/Tallerkener/Potter Laboratoriebakker/-skåle
The stainless steel bowl is suitable for a wide variety of applications, e.g. for storing, carrying or cleaning laboratory cutlery or samplers. The smooth surface of the tray makes it easy to clean.
  • For placing, transporting or cleaning instruments
  • Robust and hygienic
  • Autoclavable
The rectangular bowl has a high edge and is very stable. Ideal for use in the pharmaceutical and chemical sectors as well as the food and cosmetics sectors.