Escherichia coli WDCM 00090 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VT000902-10EA VT000904-10EA VT000905-10EA VT000906-10EA
VT000902-10EAEA 633 DKK
VT000902-10EA VT000904-10EA VT000905-10EA VT000906-10EA
Escherichia coli WDCM 00090 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Standarder Biologiske standarder
Derived from CECT® strains.
  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use
Certificeringer: These certified reference materials (CRMs) are produced and certified in accordance with ISO 17034 and ISO/IEC 17025.