Enterococcus faecalis WDCM 00009 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Leverandør: Merck
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
VT000093-10EA VT000094-10EA VT000096-10EA VT000097-10EA
VT000093-10EAEA 633 DKK
VT000093-10EA VT000094-10EA VT000096-10EA VT000097-10EA
Enterococcus faecalis WDCM 00009 Vitroids™, Certified Reference Material, Suitable for Microbiology, Supelco®
Standarder Biologiske standarder
Vitroids™ discs contain Enterococcus faecalis WDCM 00009 viable microorganisms in a certified quantity (generally accredited according to ISO/IEC 17025), produced under reproducible conditions compliant with ISO 17034:2016 using authenticated strains from CECT®.
  • Defined CFU range and low standard deviation
  • Fast, reliable and easy to use
The discs consist of pure cultures of bacteria in a solid matrix, stable for at least one year and are in a viable state with a shelf life of 1-3 years.
Each batch is provided with a comprehensive certificate of analysis that specifies the mean number of colony-forming units (CFU), an expanded uncertainty about the mean, details about the method used to determine the product data and the number of passages (subcultures) from the original strain.
Vitroids™ are derived from a traceable culture obtained freeze-dried from CECT®.
Certificeringer: This certified reference material (CRM) is produced and certified in accordance with ISO 17034 and ISO/IEC 17025.