Disposable funnel for powders, LaboPlast® / SteriPlast®
Leverandør: BURKLE
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
5378-6005 5378-6006 5379-0041 5379-1041 5378-6007 5378-6008
221-0308EA 331 DKK
221-0308 221-0309 221-0310 221-0311 221-0312 221-0313
Disposable funnel for powders, LaboPlast® / SteriPlast®
Tragte Pulvertragte
The disposable powder funnel has a short, wide discharge and is suitable for transferring powdered samples and materials, granulates and suspensions as well as other liquids.
  • For easy and reliable transferring
  • Wide opening prevents clogging
  • Suitable for use in laboratories and clean rooms or for sampling
  • Clean room manufactured (clean room Class 7)
  • Individually packaged and optionally sterilised
  • Complies with EU food requirements and FDA regulations
The wide opening of the disposable powder funnel prevents the powder or granulates from clogging while dispensing. That makes this practical and hygienic pouring aid a helpful accessory for sampling or in laboratories. In combination with a filter paper the disposable funnel is also well suited for filtering.
The sturdy disposable funnel for powdered substances is ideal for use in laboratories and clean rooms or for sampling. It can be used in pharmaceuticals, chemistry and cosmetics and the food and animal feed industries, as well as all areas where special purity is important.
Emballering: 10 pieces per pack, individually packaged.