PharmaFocus® Premium Reinforced Silicone Tubing
Leverandør: Freudenberg Medical Europe GmbH
Ratings: Ingen bedømmelse
Samlet vurdering
Gennemsntilig vurdering
49464975 49464511 49464976 49456981 49446462 49464977 49446461 49464978 49450059 49464979 49464990
HELI49464975EA 7040 DKK
HELI49464975 HELI49464511 HELI49464976 HELI49456981 HELI49446462 HELI49464977 HELI49446461 HELI49464978 HELI49450059 HELI49464979 HELI49464990
PharmaFocus® Premium Reinforced Silicone Tubing
Slanger Overførselsslange
PharmaFocus® Premium Reinforced Silicone Tubing is available with single-braid reinforcement for enhanced performance in high pressure and vacuum applications. All reinforced silicone tubing is manufactured with an inner layer of platinum-cured silicone for ultra-pure fluid transfer and meets a diverse range of pharmaceutical and bioprocessing applications.
  • Performs in high purity fluid transfer and pump applications
  • Tube resilience and adaptability due to excellent flexibility
  • High visibility of fluid transport
  • Versatile processing with cut-to-length, spools and packaging
  • Biocompatibility for demanding biomedical / pharmaceutical applications
Typical hardness: 65 Shore
Certificeringer: Material certificate and lot traceability are included with every product. Full Compliance Statements, available upon request:
> Extractables test results - BPOG & USP 665
> USP Class VI /> ISO 10993
> European Pharmacopoeia 3.1.9
> USP 85, 87, 88, 661, 788 (ASTC, ASTP)